Assay Details
| Gene Symbol: FAM198B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745288.3 | XP_002745334.1 | 3 - 4 | 1723 | 106 | |
| XM_008992543.2 | XP_008990791.1 | 3 - 4 | 1521 | 106 | ||
| XM_008992544.2 | XP_008990792.1 | 3 - 4 | 1614 | 106 | ||
More Information
Set Membership: |
|

FAM198B
Genomic Map