Assay Details
Gene Symbol: FOLR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754837.3 | XP_002754883.1 | 2 - 3 | 382 | 63 | |
XM_017977825.1 | XP_017833314.1 | 1 - 2 | 280 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
