Assay Details
Gene Symbol: PCP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008987108.2 | XP_008985356.1 | 2 - 3 | 200 | 87 | |
XM_008987109.2 | XP_008985357.1 | 2 - 3 | 188 | 87 | ||
XM_017967955.1 | XP_017823444.1 | 4 - 5 | 485 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() |
