Assay Details
Gene Symbol: CD151 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002755679.3 | XP_002755725.1 | 6 - 7 | 847 | 55 | |
GenBank mRNA | EF215096.1 | - | 2 - 3 | 106 | 55 | |
EF215392.1 | - | 2 - 3 | 111 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
