Assay Details
Gene Symbol: RBMY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267757.1 | NP_001254686.1 | 11 - 12 | 1303 | 95 | |
GenBank mRNA | FJ527002.1 | - | 11 - 12 | 1303 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
