Assay Details
| Gene Symbol: RBMY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267757.1 | NP_001254686.1 | 2 - 3 | 225 | 73 | |
| GenBank mRNA | FJ527002.1 | - | 2 - 3 | 225 | 73 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

RBMY| Gene Symbol: RBMY | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267757.1 | NP_001254686.1 | 2 - 3 | 225 | 73 | |
| GenBank mRNA | FJ527002.1 | - | 2 - 3 | 225 | 73 | |