Assay Details
Gene Symbol: RBMY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267757.1 | NP_001254686.1 | 3 - 4 | 333 | 68 | |
GenBank mRNA | FJ527002.1 | - | 3 - 4 | 333 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
