Assay Details
| Gene Symbol: WASF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750496.4 | XP_002750542.1 | 8 - 9 | 1635 | 70 | |
| XM_009000739.2 | XP_008998987.1 | 8 - 9 | 1974 | 70 | ||
More Information
Set Membership: |
|

WASF2
Genomic Map