Assay Details
| Gene Symbol: FXYD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002762009.4 | XP_002762055.1 | 4 - 5 | 336 | 98 | |
| XM_017967933.1 | XP_017823422.1 | 3 - 4 | 330 | 98 | ||
| XM_017967934.1 | XP_017823423.1 | 2 - 3 | 249 | 98 | ||
| XM_017967935.1 | XP_017823424.1 | 4 - 5 | 374 | 98 | ||
More Information
Set Membership: |
|

FXYD5
Genomic Map