Assay Details
Gene Symbol: GEMIN8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008988969.2 | XP_008987217.1 | 4 - 5 | 731 | 62 | |
XM_008988970.2 | XP_008987218.1 | 3 - 4 | 658 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
