Assay Details
Gene Symbol: PQLC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008981130.2 | XP_008979378.2 | 2 | 496 | 57 | |
XM_017964539.1 | XP_017820028.1 | 2 | 496 | 57 | ||
XM_017964540.1 | XP_017820029.1 | 5 - 6 | 496 | 57 | ||
XM_017964541.1 | XP_017820030.1 | 5 - 6 | 496 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
