Assay Details
Gene Symbol: TMEM131L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002745383.4 | XP_002745429.2 | 29 - 30 | 4094 | 66 | |
XM_008992583.2 | XP_008990831.1 | 29 - 30 | 4163 | 66 | ||
XM_008992584.2 | XP_008990832.1 | 29 - 30 | 4160 | 66 | ||
XM_008992585.2 | XP_008990833.1 | 29 - 30 | 4160 | 66 | ||
XM_008992586.2 | XP_008990834.1 | 29 - 30 | 4091 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
