Assay Details
| Gene Symbol: IL18BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754877.3 | XP_002754923.1 | 4 - 5 | 2806 | 83 | |
| XM_017977832.1 | XP_017833321.1 | 5 - 6 | 1237 | 83 | ||
| XM_017977833.1 | XP_017833322.1 | 5 - 6 | 556 | 83 | ||
| XM_017977834.1 | XP_017833323.1 | 4 - 5 | 1204 | 83 | ||
More Information
Set Membership: |
|

IL18BP
Genomic Map