Assay Details
Gene Symbol: EPHA10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017974568.1 | XP_017830057.1 | 6 - 7 | 1857 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
