Assay Details
Gene Symbol: LOC100385179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002744882.4 | XP_002744928.1 | 2 - 3 | 297 | 98 | |
XM_008991960.2 | XP_008990208.1 | 2 - 3 | 297 | 98 | ||
XM_017969874.1 | XP_017825363.1 | 2 - 3 | 208 | 98 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
