Assay Details
Gene Symbol: BIK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002743826.3 | XP_002743872.1 | 3 - 4 | 365 | 96 | |
XM_008979866.2 | XP_008978114.2 | 3 - 4 | 475 | 96 | ||
XM_017964012.1 | XP_017819501.1 | 2 - 3 | 281 | 96 |
