Assay Details
| Gene Symbol: APOBEC3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301845.1 | NP_001288774.1 | 1 | 185 | 71 | |
| GenBank mRNA | HQ404235.1 | - | 1 | 185 | 71 | |
More Information
Set Membership: |
|

APOBEC3A
Genomic Map