Assay Details
| Gene Symbol: CD40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267730.1 | NP_001254659.1 | 1 - 2 | 50 | 71 | |
| XM_008995938.1 | XP_008994186.1 | 1 - 2 | 130 | 71 | ||
| GenBank mRNA | DQ189221.1 | - | 1 - 2 | 50 | 71 | |
More Information
Set Membership: |
|

CD40
Genomic Map