Assay Details
Gene Symbol: ITM2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002749879.3 | XP_002749925.1 | 2 - 3 | 432 | 65 | |
XM_017973789.1 | XP_017829278.1 | 3 - 4 | 643 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
