Assay Details
| Gene Symbol: PPP1R16A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734981.3 | XP_003735029.2 | 1 - 2 | 959 | 55 | |
| XM_017965498.1 | XP_017820987.1 | 1 - 2 | 959 | 55 | ||
More Information
Set Membership: |
|

PPP1R16A
Genomic Map