Assay Details
| Gene Symbol: RASL10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748348.4 | XP_002748394.1 | 3 - 4 | 713 | 61 | |
| XM_008997062.2 | XP_008995310.1 | 3 - 4 | 988 | 61 | ||
More Information
Set Membership: |
|

RASL10B
Genomic Map