Assay Details
| Gene Symbol: NAA60 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003734399.3 | XP_003734447.1 | 4 - 5 | 538 | 81 | |
| XM_017978676.1 | XP_017834165.1 | 4 - 5 | 538 | 81 | ||
| XM_017978677.1 | XP_017834166.1 | 5 - 6 | 652 | 81 | ||
More Information
Set Membership: |
|

NAA60
Genomic Map