Assay Details
| Gene Symbol: FBXW4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756540.4 | XP_002756586.2 | 1 - 2 | 1574 | 72 | |
| XM_017979261.1 | XP_017834750.1 | 1 - 2 | 1574 | 72 | ||
| XM_017979262.1 | XP_017834751.1 | 1 - 2 | 1574 | 72 | ||
| XM_017979263.1 | XP_017834752.1 | 1 - 2 | 1574 | 72 | ||
More Information
Set Membership: |
|

FBXW4
Genomic Map