Assay Details
Gene Symbol: C19H7orf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008985741.2 | XP_008983989.1 | 8 | 1655 | 93 | |
GenBank mRNA | EF215113.1 | - | 1 | 418 | 93 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
