Assay Details
Gene Symbol: SPSB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_009000372.2 | XP_008998620.1 | 3 - 4 | 1522 | 66 | |
XM_017974308.1 | XP_017829797.1 | 2 - 3 | 2389 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
