Assay Details
Gene Symbol: ZNF793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001270918.1 | NP_001257847.1 | 3 - 4 | 268 | 81 | |
XM_008987872.2 | XP_008986120.1 | 8 - 9 | 891 | 81 | ||
XM_008987873.2 | XP_008986121.1 | 8 - 9 | 894 | 81 | ||
XM_017967249.1 | XP_017822738.1 | 8 - 9 | 1314 | 81 | ||
XM_017967250.1 | XP_017822739.1 | 8 - 9 | 855 | 81 | ||
XM_017967251.1 | XP_017822740.1 | 7 - 8 | 717 | 81 | ||
XM_017967252.1 | XP_017822741.1 | 9 - 10 | 1009 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
