Assay Details
| Gene Symbol: AHNAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977456.1 | XP_017832945.1 | 12 | 14155 | 101 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AHNAK2| Gene Symbol: AHNAK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017977456.1 | XP_017832945.1 | 12 | 14155 | 101 | |
Set Membership: |
|