Assay Details
| Gene Symbol: SLCO5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758977.2 | XP_002759023.1 | 3 - 4 | 1742 | 83 | |
| XM_002758978.2 | XP_002759024.1 | 3 - 4 | 1742 | 83 | ||
| XM_017965394.1 | XP_017820883.1 | 2 - 3 | 2047 | 83 | ||
More Information
Set Membership: |
|

SLCO5A1
Genomic Map