Assay Details
Gene Symbol: PLBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017976398.1 | XP_017831887.1 | 319 | 50 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
