Assay Details
Gene Symbol: C5H17orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017971685.1 | XP_017827174.1 | 2 | 223 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: C5H17orf51 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017971685.1 | XP_017827174.1 | 2 | 223 | 100 |
Set Membership: |
![]() ![]() ![]() |