Assay Details
| Gene Symbol: TBXA2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761580.3 | XP_002761626.1 | 2 - 3 | 1212 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TBXA2R| Gene Symbol: TBXA2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761580.3 | XP_002761626.1 | 2 - 3 | 1212 | 98 | |
Set Membership: |
|