Assay Details
Gene Symbol: DDX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002754495.3 | XP_002754541.1 | 3 - 4 | 714 | 107 | |
XM_009006951.2 | XP_009005199.1 | 3 - 4 | 624 | 107 | ||
XM_009006952.2 | XP_009005200.1 | 3 - 4 | 613 | 107 | ||
XM_017977594.1 | XP_017833083.1 | 3 - 4 | 650 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
