Assay Details
| Gene Symbol: CTSC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754706.3 | XP_002754752.1 | 1 - 2 | 549 | 69 | |
| XM_009007277.1 | XP_009005525.1 | 1 - 2 | 549 | 69 | ||
| XM_009007278.2 | XP_009005526.1 | 1 - 2 | 274 | 69 | ||
More Information
Set Membership: |
|

CTSC
Genomic Map