Assay Details
Gene Symbol: PRDM11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017978044.1 | XP_017833533.1 | 6 - 7 | 1869 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
