Assay Details
| Gene Symbol: LOC100399806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008999704.2 | XP_008997952.1 | 13 - 14 | 2071 | 55 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100399806| Gene Symbol: LOC100399806 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008999704.2 | XP_008997952.1 | 13 - 14 | 2071 | 55 | |
Set Membership: |
|