Assay Details
Gene Symbol: NR3C2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001267749.2 | NP_001254678.1 | 6 - 7 | 2652 | 68 | |
XM_008992564.2 | XP_008990812.1 | 7 - 8 | 3009 | 68 | ||
XM_008992565.2 | XP_008990813.1 | 7 - 8 | 2866 | 68 | ||
XM_008992566.1 | XP_008990814.1 | 7 - 8 | 2997 | 68 | ||
GenBank mRNA | AY225976.1 | - | 6 - 7 | 2642 | 68 | |
AY225977.1 | - | 6 - 7 | 2654 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
