Assay Details
Gene Symbol: LOC100412777 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002758109.4 | XP_002758155.1 | 1 - 2 | 184 | 97 | |
XM_008981310.2 | XP_008979558.1 | 1 - 2 | 184 | 97 | ||
XM_017964713.1 | XP_017820202.1 | 1 - 2 | 184 | 97 | ||
XM_017964714.1 | XP_017820203.1 | 1 - 2 | 183 | 97 | ||
XM_017964715.1 | XP_017820204.1 | 1 - 2 | 184 | 97 | ||
XM_017964716.1 | XP_017820205.1 | 1 - 2 | 184 | 97 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
