Assay Details
Gene Symbol: BICD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017976001.1 | XP_017831490.1 | 3 - 4 | 930 | 78 | |
XM_017976002.1 | XP_017831491.1 | 3 - 4 | 930 | 78 | ||
XM_017976003.1 | XP_017831492.1 | 3 - 4 | 930 | 78 | ||
XM_017976005.1 | XP_017831494.1 | 3 - 4 | 930 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
