Assay Details
Gene Symbol: PLGRKT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003731976.3 | XP_003732024.1 | 5 - 6 | 598 | 77 | |
XM_008996586.2 | XP_008994834.1 | 5 - 6 | 683 | 77 | ||
XM_008996589.2 | XP_008994837.1 | 5 - 6 | 580 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
