Assay Details
Gene Symbol: TSPAN12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752078.4 | XP_002752124.1 | 6 - 7 | 683 | 76 | |
XM_009003195.2 | XP_009001443.1 | 5 - 6 | 519 | 76 | ||
XM_009003197.2 | XP_009001445.1 | 5 - 6 | 581 | 76 | ||
XM_009003199.2 | XP_009001447.1 | 5 - 6 | 536 | 76 | ||
XM_017975714.1 | XP_017831203.1 | 5 - 6 | 719 | 76 | ||
XM_017975715.1 | XP_017831204.1 | 5 - 6 | 445 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
