Assay Details
Gene Symbol: GEMIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002757832.4 | XP_002757878.1 | 2 - 3 | 265 | 99 | |
XM_008980829.2 | XP_008979077.1 | 2 - 3 | 414 | 99 | ||
XM_017964397.1 | XP_017819886.1 | 2 - 3 | 164 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
