Assay Details
Gene Symbol: SUSD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008985160.2 | XP_008983408.1 | 8 - 9 | 1678 | 63 | |
XM_008985161.2 | XP_008983409.1 | 9 - 10 | 1689 | 63 | ||
XM_008985162.2 | XP_008983410.1 | 8 - 9 | 1672 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
