Assay Details
| Gene Symbol: LPGAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002760549.3 | XP_002760595.1 | 4 - 5 | 672 | 70 | |
| XM_017966466.1 | XP_017821955.1 | 5 - 6 | 717 | 70 | ||
| XM_017966468.1 | XP_017821957.1 | 3 - 4 | 679 | 70 | ||
More Information
Set Membership: |
|

LPGAT1
Genomic Map