Assay Details
| Gene Symbol: ZNF670 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008985654.2 | XP_008983902.1 | 3 - 4 | 438 | 109 | |
| XM_008985655.2 | XP_008983903.1 | 3 - 4 | 400 | 109 | ||
| XM_017966661.1 | XP_017822150.1 | 3 - 4 | 531 | 109 | ||
More Information
Set Membership: |
|

ZNF670
Genomic Map