Assay Details
| Gene Symbol: SYNPO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745445.3 | XP_002745491.1 | 3 - 4 | 1253 | 71 | |
| XM_008992769.2 | XP_008991017.1 | 3 - 4 | 1253 | 71 | ||
More Information
Set Membership: |
|

SYNPO2
Genomic Map