Assay Details
| Gene Symbol: MSRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755809.3 | XP_002755855.3 | 1 - 2 | 213 | 57 | |
| XM_017978646.1 | XP_017834135.1 | 1 - 2 | 213 | 57 | ||
More Information
Set Membership: |
|

MSRB1
Genomic Map