Assay Details
| Gene Symbol: IL6R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001267756.1 | NP_001254685.1 | 3 - 4 | 469 | 64 | |
| XM_008984435.2 | XP_008982683.1 | 3 - 4 | 1631 | 64 | ||
| GenBank mRNA | DQ826673.1 | - | 3 - 4 | 469 | 64 | |
More Information
Set Membership: |
|

IL6R
Genomic Map