Assay Details
Gene Symbol: HAX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002760037.4 | XP_002760083.1 | 3 - 4 | 686 | 96 | |
XM_002760038.3 | XP_002760084.1 | 3 - 4 | 697 | 96 | ||
XM_017966307.1 | XP_017821796.1 | 3 - 4 | 687 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
