Assay Details
Gene Symbol: ATF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752393.4 | XP_002752439.1 | 4 - 5 | 616 | 70 | |
XM_009003778.2 | XP_009002026.1 | 4 - 5 | 1070 | 70 | ||
XM_017976056.1 | XP_017831545.1 | 4 - 5 | 508 | 70 | ||
XM_017976057.1 | XP_017831546.1 | 4 - 5 | 442 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
