Assay Details
Gene Symbol: COQ7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002755939.4 | XP_002755985.1 | 5 - 6 | 647 | 65 | |
XM_009009244.2 | XP_009007492.1 | 5 - 6 | 529 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
